Orders LinkContactUsLinkComponent
Cianocobalamina USP
Bottle, 250 mg
  APOSBIV6236-250MG
Cianocobalamina USP
  APOSBIV6236-250MG
 :  68-19-9
Cianocobalamina USP
Bottle, 250 mg
 :  68-19-9
SFr. 26.00
1 * 250 mg
  •  :68-19-9
  •  :C₆₃H₈₈CoN₁₄O₁₄P
  •  :MFCD00151092
  •  :1355.38 g/mol
  •  :>300 °C
  •  :>300 °C
  •  :Refrigerata
  • Tipo di conf.:
    Bottle
  • Conf:
    250 mg