(S)-(-)-Hydroxypyrrolidine 97%
Bottle, 5 g
Katalognr. APOSOR4627-5G
Anbieter: Apollo Scientific
(S)-(-)-Hydroxypyrrolidine 97%
Katalognr. APOSOR4627-5G
Anbieter: Apollo Scientific
CAS Nummer: 100243-39-8
Direkt zu
Product Details & Documents
Chiral intermediate for pharmaceuticals and agrochemicals.
Synonyme: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
- CAS Nummer:100243-39-8
- Formel:C₄H₉NO
- MDL-Nummer:MFCD00192426
- Molekulargewicht:87.12 g/mol
- Siedepunkt:108…110 °C/8mmHg
- Schmelzender Platingehalt:15 °C
- Dichte:1,048 g/cm³ (25 °C)
- Blitz Pt:110 °C
- Lagertemperatur:Congelamento
Technische Daten
Technische Daten
- Tipo di conf.:Bottle
- Conf:5 g



