Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Lestaurtinib

Bottle, 1 mg
Catalog # APOSBIL2000-1MG
Irritant
Supplier:   Apollo Scientific
Lestaurtinib
Catalog # APOSBIL2000-1MG
Supplier:  Apollo Scientific
CAS Number:  111358-88-4
Lestaurtinib

Bottle, 1 mg

CAS Number:  111358-88-4
Price
SFr. 123.00
Log in for availability
1 * 1 mg
Product Details & Documents
  • Protein
  • CAS Number:111358-88-4
  • Storage Temperature:Congelamento
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    1 mg

Specifications