EDTA ≥99%
Bottle, 500 g
Codice catalogo APOSBIE0729-500G
Nome del fornitore: Apollo Scientific
Vai a
Product Details & Documents
Sinonimi: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- Numero CAS:60-00-4
- Formula:C₁₀H₁₆N₂O₈
- Numero MDL:MFCD00003541
- Molecular Weight:292.25 g/mol
- Punto di fusione:245 °C (dec.)
- Densità:1,46 g/cm³ (25 °C)
- temperatura di conservazione:Ambiente
- EINECS:200-449-4
Specifiche
Specifiche
- Tipo di conf.:Bottle
- Conf:500 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



