Glicina ≥99%
Bottle, 1 kg
Catalog # APOSBIG0709-1KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.
- Ph. Eur.
- Pyrogen free
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Melting Pt:240…245 °C (dec.)
- Density:1,595 g/cm³ (25 °C)
- Storage Temperature:Ambiente
- EINECS:200-272-2
Specifications
Specifications
- Tipo di conf.:Bottle
- Conf:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



