Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cable ties, non-releasable

Cable ties, non-releasable

rosso, 3,5×200 mm
N° de catalogue BRDY196351
Fournisseur:   Brady
Cable ties, non-releasable
N° de catalogue BRDY196351
Fournisseur:  Brady
Cable ties, non-releasable
Cable ties, non-releasable

rosso, 3,5×200 mm

Prix
SFr. 13.20
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

Nylon cable ties, easily fixed around items up to 165 mm in circumference.

Ideal for colour-coding hard to mark items.

Spécifications
  • Colore:
    rosso
  • Dimensioni:
    3,5×200 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.