Blind cap
29/32 NS
Katalognr. NEUB1670-10-29
Anbieter: NEUBERT VOLUME GLASSWAERE
Direkt zu
Product Details & Documents
Ground cap with standard ground joints made of borosilicate glass 3.3.
- Very good chemical resistance
- Very good temperature resistance
Technische Daten
- Dimensione cono:29/32 NS
- Seq:02