Acebutolol, Reference Standards
Acebutolol, USP reference standard, 20 mg
Codice catalogo 1000699.
Nome del fornitore: USP
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Conf:20 mg
- Standard:USP reference standard
- Descrizione:Acebutolol
- Cat. No.:1000699.
- Descrizione:Acebutolol
- Synonym(s):N-{3-Acetyl-4-[3-(ethylamino)-2-hydroxypropoxy]phenyl}butyramide
- CAS No.:441019-91-6
Specifiche
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



