Human B7-1, His Tag (c-terminus) is expressed from human 293 cells. It contains AA Val 35 - Asn 242 (Accession # NP_005182.1) 1 * 100 µG
Catalog # ACRBB71-H5228100UG
Supplier: ACRO Biosystems
Human B7-1, His Tag (c-terminus) is expressed from human 293 cells. It contains AA Val 35 - Asn 242 (Accession # NP_005182.1) 1 * 100 µG
Catalog # ACRBB71-H5228100UG
Supplier: ACRO Biosystems
Frequently Bought Together
[EN]DIMETHYL SULFOXIDE EMPLURA 1 * 185 L
Supplier: Merck
Dibromometano stabilizzato per sintesi, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Metanolo ≥99.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Etanolo denaturato ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Cloroformio 99.0-99.4% (by GC) stabilizzato, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Cesio cloruro 99.99% (trace metals basis), Optical Grade per spettroscopia
Supplier: Thermo Fisher Scientific
Dimetilsolfossido ≥99.9% (by GC), CHROMASOLV™ per cromatografia di spazio di testa, Riedel-de Haën™
Supplier: Honeywell Chemicals
ter-Butanolo ≥99%
Supplier: Thermo Fisher Scientific
n-Pentano 99+%, extra puro
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Metanolo ACS, grado gradiente per HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
1,2-Diclorobenzene 99+% per HPLC
Supplier: Thermo Fisher Scientific
n-Esano 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Supplier: PanReac AppliChem