Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Qualitix® Pipette Tips

Ultra-micro tips, fine point, 10 µl, -, 31,4 mm, Trasparente, 12 racks, 96 each
  SOCO3090.0010R
Qualitix® Pipette Tips
  SOCO3090.0010R
Qualitix® Pipette Tips
Qualitix® Pipette Tips

Ultra-micro tips, fine point, 10 µl, -, 31,4 mm, Trasparente, 12 racks, 96 each

 
148.00 SFr.
   
1 * 1.152 ST
 

These tips are made of 100% pure virgin, medical grade PP, fitting not only Socorex® but all major micropipette brands.

  • Superior quality, autoclavable
  • Lot by lot consistency
  • Certified free of nuclease, endotoxin and ATP
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with other micropipette brands

Beveled tips: Special shape eliminates liquid drop sticking at tip end.
Fine pointed tips: Have a thin, soft end enabling smooth liquid deposit.

Low binding tips are made with a silicone-free resin reducing surface adhesion. They provide maximal liquid recovery. Best suited for proteomic research and handling of any viscous liquids. Protease-free certified.

 : The tipfill system is intended for easy rack loading. Tip trays are instantly transferred into a rack without risk of contamination. The smart re-closable tipfill system box reduces storage space, waste volume and shipping costs.

 
  • Confezione:
    12 racks, 96 each
  • Sterili:
    -
  • Volume:
    10 µl
  • Lunghezza:
    31,4 mm
  • Seq:
    04
  • Stile punta:
    Ultra-micro tips, fine point
  • Colore puntale:
    Trasparente
Accessories

 

  •  
    Descrizione
     
     
  • SOCO3070.0210ER
    Empty rack for 10/200 µl tips, to be used with tipfill system
       
    1 * 10 ST - SFr. 57.30

 

  •  
    Descrizione
     
     
  • SOCO3070.1000ER
    Empty rack for 1000 µl tips, to be used with tipfill system
       
    1 * 6 ST - SFr. 49.00