Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Conductivity standards, at 25 °C, Supelco®

Conductivity standard, 100000 μ S/cm, 25 °C, 500 mL
  59777-500ML
 :   Merck
Conductivity standards, at 25 °C, Supelco®
  59777-500ML
 :  Merck
 :  
Conductivity standards, at 25 °C, Supelco®

Conductivity standard, 100000 μ S/cm, 25 °C, 500 mL

 :  
 
SFr. 232.00
   
1 * 500 mL
 

 

 

 

  • Conf:
    500 mL
  • Temperatura di riferimento:
    25 °C
  • Descrizione:
    Conductivity standard
  • Conduttanza:
    100000 μ S/cm