Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
BOD calibration standards, (CRM), Supelco®

BOD calibration standards, (CRM), Supelco®

BOD calibration standard, Certified reference material, 500 mg/l in water, 500 mL
Catalog # BOD500-500ML
Supplier:   Merck
BOD calibration standards, (CRM), Supelco®
Catalog # BOD500-500ML
Supplier:  Merck
CAS Number:  
BOD calibration standards, (CRM), Supelco®

BOD calibration standard, Certified reference material, 500 mg/l in water, 500 mL

CAS Number:  
Price
SFr. 98.70
Log in for availability
1 * 500 mL
Product Details & Documents

Certifications: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.

Caution: Storage temperature: 2 - 8 °C (refrigerator)

Documents

Specifications

Specifications

  • Concentrazione:
    500 mg/l in water
  • Conf:
    500 mL
  • Descrizione:
    BOD calibration standard
  • Tipo:
    Certified reference material

Specifications