BOD calibration standards, (CRM), Supelco®
BOD calibration standard, Certified reference material, 200 mg/l in water, 100 mL
Codice catalogo BOD200-100ML
Nome del fornitore: Merck
BOD calibration standards, (CRM), Supelco®
Codice catalogo BOD200-100ML
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
Certifications: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Attenzione: Storage temperature: 2 - 8 °C (refrigerator)
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Concentrazione:200 mg/l in water
- Conf:100 mL
- Descrizione:BOD calibration standard
- Tipo:Certified reference material
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



