BOD calibration standards, (CRM), Supelco®
BOD calibration standard, Certified reference material, 2000 mg/l in water, 500 mL
Nome del fornitore: Merck
BOD calibration standards, (CRM), Supelco®
Codice catalogo BOD2000-500ML
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
: Storage temperature: 2 - 8 °C (refrigerator)
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Concentrazione:2000 mg/l in water
- Conf:500 mL
- Descrizione:BOD calibration standard
- Tipo:Certified reference material
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



