Specific Gravity Hydrometers
Specific gravity hydrometer, 106 H - 62, 0,850 - 0,900 °SG, 0,001 °SG, 260 mm
Katalognr. AMARH826788DKD3
Anbieter: Amarell
Direkt zu
Product Details & Documents
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
- Capillary prismatic
- Reliable and simple
- Paper scale with type designation
- Shot weighted body
Informazioni per l'ordine: Supplied in a transparent plastic case.
Technische Daten
- Descrizione:Specific gravity hydrometer
- Lunghezza:260 mm
- Seq:48
- Divisione:0,001 °SG
- Modello:106 H - 62
- Range di misura:0,850 - 0,900 °SG
- Cat. No.:AMARH826788DKD3
- Temperatura di riferimento:60/60 °F (except model AMARH826764-FL: 15 °C)
- Accuratezza:±1 scale division