Inserts for autosampler vials, AQ™
Insert for 4 ml (15×45 mm) screw top vials, deactivated silanised, 0,70 ml, Precision point, 8×40 mm, Vetro borosilicato 3.3, No
Katalognr. MICS9504S-03ND
Anbieter: MICROSOLV
Direkt zu
Product Details & Documents
These vial inserts are made from borosilicate glass or PP.
Technische Daten
- Tappo incluso:No
- Ø×A:8×40 mm
- Capacità:0,70 ml
- Descrizione:Insert for 4 ml (15×45 mm) screw top vials, deactivated silanised
- Materiale:Vetro borosilicato 3.3
- Seq:28
- Fondo:Precision point