Inserts for autosampler vials, AQ™
Insert for 2 ml vials, deactivated silanised, 0,25 ml, Conical bottom, 6×31 mm, Vetro borosilicato, No
Catalog # MICS9502S-03ND
Supplier: MICROSOLV
Jump to:
Product Details & Documents
These vial inserts are made from borosilicate glass or PP.
Specifications
- Tappo incluso:No
- Ø×A:6×31 mm
- Capacità:0,25 ml
- Descrizione:Insert for 2 ml vials, deactivated silanised
- Materiale:Vetro borosilicato
- Seq:16
- Fondo:Conical bottom