Drum faucets
Internal flame arrester, adjustable, 3/4"
Codice catalogo 119-0240
Nome del fornitore: JUSTRITE MANUFACTURING
Vai a
Product Details & Documents
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
- Safe and easy dispensing
- FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
Specifiche
- Filettatura:3/4"
- Descrizione:Internal flame arrester, adjustable
- Seq:02
- Cat. No.:119-0240