Orders LinkContactUsLinkComponent
Double spatulas, bent
Double spatulas, bent
250 mm, 12 mm
Catalog # USBE3295
Supplier:   USBECK Laborgeräte
Double spatulas, bent
Catalog # USBE3295
Double spatulas, bent
Double spatulas, bent
250 mm, 12 mm
Price
SFr. 5.10
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
Specifications
  • Lunghezza:
    250 mm
  • Seq:
    04
  • Larghezza lama:
    12 mm