Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Disposable knives, SteriWare®

Disposable knives, SteriWare®

Disposable knife, non sterile, individually bagged, 60 mm, 160 mm, bianco
  SAMP8039H-100
Disposable knives, SteriWare®
  SAMP8039H-100
Disposable knives, SteriWare®
Disposable knives, SteriWare®

Disposable knife, non sterile, individually bagged, 60 mm, 160 mm, bianco

 
SFr. 82.90
   
1 * 100 ST
 

These single use knives are made from food grade plastic (100% virgin HDPE). Ideal for sample preparation and general laboratory use.

  • Can be used to cut or split products
  • Available non sterile or gamma irradiated sterile (SAL 10-6)
  • Moulded and packed in a cleanroom
  • No cleaning is required before or after sampling
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • BSE/TSE-free

Certifications: Material conforms to FDA and EU 10/2011. Full batch traceability.

 
  • Colore:
    bianco
  • Descrizione:
    Disposable knife, non sterile, individually bagged
  • Seq:
    02
  • Lunghezza totale:
    160 mm
  • Lama:
    60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.