Drying chambers
Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
Codice catalogo LBCP7318800
Nome del fornitore: LABCONCO
Vai a
Product Details & Documents
Specifiche
- Descrizione:Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
- Seq:02
- Cat. No.:LBCP7318800
- Environmentally Preferable:Imballi sostenibili