Orders LinkContactUsLinkComponent
Drying chambers
Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
Catalog # LBCP7318802
   Imballi sostenibili
Supplier:   LABCONCO
Drying chambers
Catalog # LBCP7318802
Supplier:  LABCONCO
   Imballi sostenibili
Drying chambers
Drying chambers
Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
Price
SFr. 3’900.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Descrizione:
    Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
  • Seq:
    03
  • Cat. No.:
    LBCP7318802
  • Environmentally Preferable:
    Imballi sostenibili