Drying chambers
Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
Catalog # LBCP7318802
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Descrizione:Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
- Seq:03
- Cat. No.:LBCP7318802
- Environmentally Preferable:Imballi sostenibili