Connecting adapters, with parallel necks
18,8/26 NS, 18,8/26 NS
Catalog # NEUB0900-10-191929
Supplier: NEUBERT VOLUME GLASSWAERE
Connecting adapters, with parallel necks
Catalog # NEUB0900-10-191929
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Two necked adapters made of borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifications
- Dimensione cono:18,8/26 NS
- Cono femmina:18,8/26 NS
- Seq:02