ContactUsLinkComponent
Human IgG Fc, Tag Free (FCC-H5214) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession P01857-1). 1 * 1 mg
Codice catalogo ACRBFCC-H5214-1MG
Nome del fornitore:  ACROBIOSYSTEMS
Human IgG Fc, Tag Free (FCC-H5214) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession P01857-1). 1 * 1 mg
Codice catalogo ACRBFCC-H5214-1MG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together