Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant IL-6

Human Recombinant IL-6

20 µG
Codice catalogo STMC78050.1
Marketsource
Nome del fornitore:   Stemcell Technologies
Human Recombinant IL-6
Codice catalogo STMC78050.1
Nome del fornitore:  Stemcell Technologies
Marketsource
Numero CAS:  
Human Recombinant IL-6
Human Recombinant IL-6

20 µG

Numero CAS:  
Price
SFr. 210.00
Log in per disponibilità
1 * 20 µG
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Interleukin 6 (IL-6) is a pleiotropic growth factor with the wide range of biological activities in immune regulation, hematopoiesis, and oncogenesis. IL-6 is produced by a variety of cell types including T cells, B cells, monocytes and macrophages, fibroblasts, hepatocytes, vascular endothelial cells, and various tumor cell lines. On its own or in combination with other factors such as IL-2 and interferon-γ, IL-6 stimulates the proliferation of B cells, T cells, and hybridoma cells (Hirano et al.; Mihara et al.; Tanaka et al). In combination with cytokines such as IL-3, GM-CSF and SCF, IL-6 has been shown to promote hematopoietic progenitor cell proliferation and differentiation in vitro. IL-6 signals through a cell surface type I cytokine receptor complex consisting of the ligand-binding IL-6α (CD126) and the signal-transducing gp130 subunits. The binding of IL-6 to its receptor system includes activation of JAK/STAT signaling pathway (Mihara et al.; Peters et al.; Tanaka et al.).

Attenzione: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifiche

Specifiche

  • Conf:
    20 µG
  • Cat. No.:
    STMC78050.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-6
  • Purezza:
    ≥95%
  • Specie:
    Human

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..