Auftragserfassung
Switzerland
ContactUsLinkComponent
CUSTOM BOX OF 48 PHYTIP COLUMNS (500 µL VOLUME) CONTAINING 40 µL OF MAIIA EPO PURIFICATION GEL (CATALOG#1271) PER COLUMN FOR TECAN FREDOM EVO MCA 96- WELL PLATFORMS 1 * 1 KIT
  BITAPTM45-40-MY
 :  BIOTAGE
CUSTOM BOX OF 48 PHYTIP COLUMNS (500 µL VOLUME) CONTAINING 40 µL OF MAIIA EPO PURIFICATION GEL (CATALOG#1271) PER COLUMN FOR TECAN FREDOM EVO MCA 96- WELL PLATFORMS 1 * 1 KIT
  BITAPTM45-40-MY
 :  BIOTAGE
Frequently Bought Together