Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Shield™ DNA/RNA Reagent

Shield™ DNA/RNA Reagent

DNA/RNA Shield™ (2X Concentrate), 125 ml, 125 mL
Catalog # ZYMOR1200-125
Marketsource
Supplier:   Zymo Research
Shield™ DNA/RNA Reagent
Catalog # ZYMOR1200-125
Supplier:  Zymo Research
Marketsource
CAS Number:  
Shield™ DNA/RNA Reagent
Shield™ DNA/RNA Reagent

DNA/RNA Shield™ (2X Concentrate), 125 ml, 125 mL

CAS Number:  
Price
315.00 SFr.
Log in for availability
1 * 125 mL
Product Details & Documents

DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.

  • Nucleic acid preservation (at ambient temperature; cold-free)
  • Pathogen inactivation (bacteria, fungus, parasites & viruses)
  • RNA Stability: Ambient temperature (4 to 25 °C) >1 month
  • DNA Stability: Ambient temperature (4 to 25 °C) >2 years
  • DNA and RNA: Frozen (<–20 °C): Indefinitely

DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialised equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).

Specifications

Specifications

  • Conf:
    125 mL
  • Descrizione:
    DNA/RNA Shield™ (2X Concentrate)
  • Volume:
    125 ml
  • Cat. No.:
    ZYMOR1200-125
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA|Total DNA
  • Target molecule:
    Copurification of DNA & RNA|DNA|RNA
  • Method / Format:
    Chemical extraction & preservation
  • Sample type:
    Stool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Descrizione:
    DNA/RNA Shield™ (2X Concentrate)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.