Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Quick-DNA™ Plus Kits

Quick-DNA™ Plus Kits

Quick-DNA™ 96 Plus kit, 2×96 Preps, 1 KIT
  ZYMOD4070
Marketsource
Quick-DNA™ Plus Kits
  ZYMOD4070
Marketsource
 :  
Quick-DNA™ Plus Kits
Quick-DNA™ Plus Kits

Quick-DNA™ 96 Plus kit, 2×96 Preps, 1 KIT

 :  
 
SFr. 808.00
   
1 * 1 KIT
 

DNA from any sample (cells, tissues, biological fluids and more).

  • Quick and easy: Simple 20 minute procedure
  • Highest yield: Recover 3× more DNA
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA >50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.

 

 

  • Conf:
    1 KIT
  • Descrizione:
    Quick-DNA™ 96 Plus kit
  • Volume:
    2×96 Preps
  • Cat. No.:
    ZYMOD4070
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Multiwell plate
  • Sample type:
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Resa:
    The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • Descrizione:
    Quick-DNA™ 96 Plus kit
  • Capacità campioni:
    5 µg
  • Dimensione del campione:
    ≤1×10⁶ Cells or 5 mg Tissue

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.