Quick-DNA™ Plus Kits
Quick-DNA™ Miniprep Plus kit, 50 Preps, 1 KIT
Codice catalogo ZYMOD4068
Marketsource
Nome del fornitore: Zymo Research
Quick-DNA™ Plus Kits
Codice catalogo ZYMOD4068
Nome del fornitore: Zymo Research
Marketsource
Numero CAS:
Vai a
Product Details & Documents
DNA from any sample (cells, tissues, biological fluids and more).
- Quick and easy: Simple 20 minute procedure
- Highest yield: Recover 3× more DNA
- Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more
The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA >50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.
Specifiche
Specifiche
- Conf:1 KIT
- Descrizione:Quick-DNA™ Miniprep Plus kit
- Volume:50 Preps
- Cat. No.:ZYMOD4068
- Prep size:Mini
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Column (silica-based)
- Sample type:Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Resa:The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
- Descrizione:Quick-DNA™ Miniprep Plus kit
- Capacità campioni:25 µg
- Dimensione del campione:≤5×10⁶ Cells or 25 mg Tissue



