Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Laboratory bottles, VITgrip™

1000 ml, GL 45, 234 mm, 97 mm
  VITL110494
 :   VITLAB
Laboratory bottles, VITgrip™
  VITL110494
 :  VITLAB
Laboratory bottles, VITgrip™
Laboratory bottles, VITgrip™

1000 ml, GL 45, 234 mm, 97 mm

 
SFr. 117.00
   
1 * 6 ST
 

Made of PP, translucent, with tamper-evident closures.

  • With graduation
  • Excellent handling
  • Leakproof and break resistant
  • Safe storage
  • Robust and very versatile
  • Excellent chemical and temperature resistance

The laboratory bottle VITgrip™ is the allround bottle for storage and sample collection of liquids in the lab. The innovative design is characterised, amongst other things, by optimum grip and high accuracy of the graduation (±5%) and provides safe protection against leakage of liquids.

 : Suitable for contact with foodstuffs (according to regulation EU No. 10/2011).

 
  • Ø est:
    97 mm
  • Filettatura:
    GL 45
  • Capacità:
    1000 ml
  • Altezza:
    234 mm
  • Seq:
    04