ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 7.3 MG/MLITRACONAZOLE IS A TRIAZOLE ANTIFUNGAL AGENT.IC50 VALUE: N/ATAR GET: ANTIFUNGALIN VITRO: ITRACONAZOLE IS PHARMACOLOGICALLY DISTINCT FROM OTHER AZOLE ANTIFUNGAL AGENTS IN THAT IT IS 1 * 500 mg
Codice catalogo MDEXHY-17514-500MG
Nome del fornitore:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 7.3 MG/MLITRACONAZOLE IS A TRIAZOLE ANTIFUNGAL AGENT.IC50 VALUE: N/ATAR GET: ANTIFUNGALIN VITRO: ITRACONAZOLE IS PHARMACOLOGICALLY DISTINCT FROM OTHER AZOLE ANTIFUNGAL AGENTS IN THAT IT IS 1 * 500 mg
Codice catalogo MDEXHY-17514-500MG
Nome del fornitore:  MEDCHEMEXPRESS
Frequently Bought Together