ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: SOLUBLE TO 100 MM IN WATER AND TO 100 MM IN DMSOA 419259 3HCL IS AN APOPTOSIS I NDUCING AGENT THAT INHIBITS SRC FAMILY KINASES (C-SRC). A 419259 ALSO INDUCES APOPTOSIS IN CML CELL INES AND BLOCKS S 1 * 10 mg
Codice catalogo MDEXHY-15764A-10MG
Nome del fornitore:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: SOLUBLE TO 100 MM IN WATER AND TO 100 MM IN DMSOA 419259 3HCL IS AN APOPTOSIS I NDUCING AGENT THAT INHIBITS SRC FAMILY KINASES (C-SRC). A 419259 ALSO INDUCES APOPTOSIS IN CML CELL INES AND BLOCKS S 1 * 10 mg
Codice catalogo MDEXHY-15764A-10MG
Nome del fornitore:  MEDCHEMEXPRESS
Frequently Bought Together