ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: SOLUBLE TO 100 MM IN DMSOTCS JNK 5A(JNK INHIBITOR IX) IS A SELECTIVE INHIBITOR OF JNK2 AND JNK3 (PIC50 VALUES ARE 6.7, 6.5, <5.0 AND <4.8 FOR JNK3, JNK2, JNK1 AND P38Α RESPECTIVELY) DISPLAYS NO SI 1 * 10 mg
Codice catalogo MDEXHY-15881-10MG
Nome del fornitore:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: SOLUBLE TO 100 MM IN DMSOTCS JNK 5A(JNK INHIBITOR IX) IS A SELECTIVE INHIBITOR OF JNK2 AND JNK3 (PIC50 VALUES ARE 6.7, 6.5, <5.0 AND <4.8 FOR JNK3, JNK2, JNK1 AND P38Α RESPECTIVELY) DISPLAYS NO SI 1 * 10 mg
Codice catalogo MDEXHY-15881-10MG
Nome del fornitore:  MEDCHEMEXPRESS
Frequently Bought Together