LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 100 MM IN DMSOIWR-1(IWR-1-ENDO) IS A NOVEL SMALL-MOLECULE INHIBITOR OF WNT SIGN ALING BY STABILIZING THE AXIN DESTRUCTION COMPLEX WITH AN EC50 OF 0.2 UM.IC50 VALUE: 0.2 UM (EC50) [1]TARGET: WNT SIG 1 * 5 mg
Codice catalogo MDEXHY-12238-5MG
Nome del fornitore: MEDCHEMEXPRESS
Frequently Bought Together