LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOSIMEPREVIR (TMC 435350) IS A POTENT INHIBITOR OF HCV NS3/4A PROTEA SE (KI = 0.36 NM) AND VIRAL REPLICATION (REPLICON EC50 = 7.8 NM). IC50 VALUE: 0.36 NM(KI) 7.8 NM (EC50) [1]TARGET: H 1 * 1 mL
Codice catalogo MDEXHY-10241-1ML
Nome del fornitore: MEDCHEMEXPRESS
Frequently Bought Together