ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSODMXAA IS A COMPETITIVE INHIBITOR OF THE ENZYME DT-DIAPHORASE (NAD( P)H:QUINONE OXIDOREDUCTASE EC 1.6.99.2) WITH RESPECT TO NADH, WITH KI VALUES OF 20 ΜM. [1]IC50 VALUE: 20 ΜM (KI) [1]T 1 * 100 mg
Catalog # MDEXHY-10964-100MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSODMXAA IS A COMPETITIVE INHIBITOR OF THE ENZYME DT-DIAPHORASE (NAD( P)H:QUINONE OXIDOREDUCTASE EC 1.6.99.2) WITH RESPECT TO NADH, WITH KI VALUES OF 20 ΜM. [1]IC50 VALUE: 20 ΜM (KI) [1]T 1 * 100 mg
Catalog # MDEXHY-10964-100MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together