Labels for BBP33 printers, type B-424
Printer labels for general identification, B-424, B33-20-424, Paper, Permanente, a base di gomma, Thermal transfer, bianco, BBP33, 76,2×127 mm
BRDY133832
: Brady
These paper labels are used for general identification and have a matt finish.
- Stampante:BBP33
- Printing method:Thermal transfer
- Tipo:B-424
- Colore:bianco
- Descrizione:Printer labels for general identification
- Dimensioni:76,2×127 mm
- Materiale:Paper
- Seq:07
- Modello:B33-20-424
- Adesivo:Permanente, a base di gomma