Orders LinkContactUsLinkComponent
Yeast Extract Agar
Yeast extract agar, 20 ml, Tube, 1
  LIOF34074
 :   LIOFILCHEM
Yeast Extract Agar
  LIOF34074
 :  
Yeast Extract Agar
Yeast extract agar, 20 ml, Tube, 1
 :  
SFr. 78.60
1 * 1 ST
Medium for microorganisms enumeration in water.
Certifications: ISO6222 certified.
  • Tipo di conf.:
    Tube
  • Conf:
    1
  • Descrizione:
    Yeast extract agar
  • Volume:
    20 ml