[EN]SNAP-ON CAP 11MM RED SILICONE/PTFE C 1 * 100 ST
Catalog # BROW153346
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
[EN]SNAP-ON CAP 11MM RED SILICONE/PTFE C 1 * 100 ST
Catalog # BROW153346
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ammonio acetato ≥99%, HiPerSolv CHROMANORM® per LC-MS
Supplier: VWR Chemicals
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Metanolo ≥99.8%, HiPerSolv CHROMANORM®, grado isocratico per HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Snap-Cap, 11mm, Farbe : ROT Silicone/PTFE Septum, 100 Stk. OMNILAB 0 1 * 100 ST
Supplier: BROWN CHROMATOGRAPHY SUPPLIES