Sedie e sgabelli da laboratorio Labsit
Black artificial leather and white backrest, 520 - 770 mm, Glides, +
Catalog # 132-1279
Supplier: bimos
Jump to:
Product Details & Documents
Labsit chairs are perfectly suited for laboratory work. Labsit offers outstanding comfort, laboratory ergonomics and intuitive operation.
Certifications:
Accessories information:
Ordering information:
Delivery information:
Packaging:
Caution:
Specifications
- Descrizione:Black artificial leather and white backrest
- Altezza:520 - 770 mm
- Seq:26
- Base:Glides
- Poggiapiedi:+