Orders LinkContactUsLinkComponent
Blocks for digital heating/cooling block heater
Interchangeable block, 50 ml centrifuge tubes (small), 6, 98×76,5×51 mm
Codice catalogo BARN88880132
Nome del fornitore:   Thermo Fisher Scientific
Blocks for digital heating/cooling block heater
Codice catalogo BARN88880132
Nome del fornitore:  Thermo Fisher Scientific
Blocks for digital heating/cooling block heater
Interchangeable block, 50 ml centrifuge tubes (small), 6, 98×76,5×51 mm
Price
288.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Blocks for digital block heater to optimise flexibility and performance.
Specifiche
  • N. di fori:
    6
  • Descrizione:
    Interchangeable block
  • Seq:
    03
  • L×P×A:
    98×76,5×51 mm
  • Per:
    50 ml centrifuge tubes (small)