Standard hazard warning signs
Warning - corrosive substances (W023), In PE, 25×22 mm
Codice catalogo BRDY829719
Nome del fornitore: Brady
Vai a
Product Details & Documents
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
- Used for a wide range of applications
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Thickness: 1,5 mm
- Non-adhesive
Specifiche
- Descrizione:Warning - corrosive substances (W023)
- Dimensioni:25×22 mm
- Materiale:In PE
- Seq:08



