Orders LinkContactUsLinkComponent
Labels, energy source, electrical, Brady
Energy source labels, E-15-SA/5, Poliestere, bianco/rosso, 75×38 mm
  BRDY138823
 :   Brady
Labels, energy source, electrical, Brady
  BRDY138823
 :  Brady
Labels, energy source, electrical, Brady
Labels, energy source, electrical, Brady
Energy source labels, E-15-SA/5, Poliestere, bianco/rosso, 75×38 mm
SFr. 23.50
1 * 5 ST
These electrical energy source labels are made from laminated polyester.
  • Permanent acrylic adhesive
  • Two different sizes available
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
 : ATEX
 : 100 labels per pack or 5 labels per pack.
  • Colore:
    bianco/rosso
  • Descrizione:
    Energy source labels
  • Dimensioni:
    75×38 mm
  • Materiale:
    Poliestere
  • Seq:
    35
  • Modello:
    E-15-SA/5