Orders LinkContactUsLinkComponent
Labels, energy source, electrical, Brady | Avantor
Labels, energy source, electrical, Brady
Energy source labels, E-9-19x13mm-SA/100, Poliestere, bianco/rosso, 19×13 mm
  BRDY138460
 :   Brady
Labels, energy source, electrical, Brady
  BRDY138460
 :  Brady
Labels, energy source, electrical, Brady
Labels, energy source, electrical, Brady
Energy source labels, E-9-19x13mm-SA/100, Poliestere, bianco/rosso, 19×13 mm
SFr. 23.50
1 * 100 ST
These electrical energy source labels are made from laminated polyester.
  • Permanent acrylic adhesive
  • Two different sizes available
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Certifications: ATEX
Confezione: 100 labels per pack or 5 labels per pack.
  • Colore:
    bianco/rosso
  • Descrizione:
    Energy source labels
  • Dimensioni:
    19×13 mm
  • Materiale:
    Poliestere
  • Seq:
    09
  • Modello:
    E-9-19x13mm-SA/100