Labels, energy source, electrical, Brady
Energy source labels, E-5-19x13mm-SA/100, Poliestere, bianco/rosso, 19×13 mm
Codice catalogo BRDY138456
Nome del fornitore: Brady
Vai a
Product Details & Documents
These electrical energy source labels are made from laminated polyester.
- Permanent acrylic adhesive
- Two different sizes available
- Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Certifications: ATEX
Packaging: 100 labels per pack or 5 labels per pack.
Specifiche
- Colore:bianco/rosso
- Descrizione:Energy source labels
- Dimensioni:19×13 mm
- Materiale:Poliestere
- Seq:05
- Modello:E-5-19x13mm-SA/100