Consignment kit
Basic consignment kit, bianco
Catalog # BRDY806190
Supplier: Brady
Jump to:
Product Details & Documents
Adjustable consignment kit used to condense flywheel valves, ball valves, butterfly valves and other valves.
Small adjustable consignment kit consists of large consignment pouch, adjustable steering valve device, SUCC with armored cable, security locks (2), 25 mm red locking devices (2), small basic fastening device for condemnation of valves, small locking arm, pen, 'Do not touch' labels (75×160 mm).
Basic consignment kit consists of small locking arm, cable tie, SUCC with armored cable, 1 security lock box, white label (75×160 mm), pen, large consignment pouch.
Specifications
- Tipo:Basic consignment kit
- Colore:bianco
- Seq:02