ContactUsLinkComponent
ART PHOTONICS LWL Kabel NIR600/660-3500-SMA-SMA-PVC55+G, Fiber: NIRsilica/silica, core 600um clad 660um, NA=0.22+-0.02, Length: 35+-1m,Termination 1: SMA 905 connector (hexagonal nut), extens 1 * 1 ST
  MSPPAP11269
ART PHOTONICS LWL Kabel NIR600/660-3500-SMA-SMA-PVC55+G, Fiber: NIRsilica/silica, core 600um clad 660um, NA=0.22+-0.02, Length: 35+-1m,Termination 1: SMA 905 connector (hexagonal nut), extens 1 * 1 ST
  MSPPAP11269
Frequently Bought Together