Orders LinkContactUsLinkComponent
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™ ACF dendritic cell medium, 500 mL
Codice catalogo STMC10986
Marketsource
Nome del fornitore:   Stemcell Technologies
ImmunoCult™-ACF Dendritic Cell Medium
Codice catalogo STMC10986
Nome del fornitore:  Stemcell Technologies
Marketsource
Numero CAS:  
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™ ACF dendritic cell medium, 500 mL
Numero CAS:  
Price
SFr. 457.13
Log in per disponibilità
1 * 500 mL
Product Details & Documents
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation.
Reliably culture and differentiate human monocytes into dendritic cells (DCs) using the serum- and animal component-free (ACF) ImmunoCult™-ACF Dendritic Cell Medium. For your convenience, the medium is available either individually or as part of ImmunoCult™ Dendritic Cell Culture Kit. Use the ImmunoCult™-ACF Dendritic Cell Medium in combination with ImmunoCult™-ACF Dendritic Cell Differentiation Supplement to generate DCs in vitro.
To support the maturation of immature DCs to mature DCs, combine with the ImmunoCult™ Dendritic Cell Maturation Supplement.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifiche
Specifiche
  • Conf:
    500 mL
  • Descrizione:
    ImmunoCult™ ACF dendritic cell medium