Orders LinkContactUsLinkComponent
Flex bench family, InfinityLab
Flex bench family, InfinityLab
Power cord cable assembly, 18-AWG, 3-conductor
Codice catalogo HEWL8121-2258
Nome del fornitore:   AGILENT HEWLETT PACKARD
Flex bench family, InfinityLab
Codice catalogo HEWL8121-2258
Nome del fornitore:  AGILENT HEWLETT PACKARD
Flex bench family, InfinityLab
Power cord cable assembly, 18-AWG, 3-conductor
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The InfinityLab flex bench family includes the InfinityLab flex bench system and the InfinityLab benchtop system.
  • Modify the height of your LC instrument, for easy access to solvent bottles and increased instrument optimisation
  • Safely configure your instrument to suit the way you work
  • Get more flexibility in how to set up your LC system - The InfinityLab benchtop system is engineered to provide an adjustable and safe way to optimise space and to configure your LC system to suit your needs for higher productivity
The InfinityLab flex bench family provides easy ways for you to configure your instrument to suit your needs, anywhere in the lab. These three sturdy steel benches are all adjustable, enabling easy and safe access to instrument components, solvent bottles, pumps, columns, and accessories. InfinityLab flex bench: move your instruments where you need them, when you need them. Whether you want to work next to your MS, or move analysis to new projects, the InfinityLab flex bench enables space optimisation while taking the hassle out of instrument relocation. InfinityLab benchtop: easily change modules and perform maintenance with your existing bench setup. The InfinityLab benchtop fits into your existing lab setup, while providing an easy way to configure your LC system. InfinityLab flex bench MS: optimise your whole lab set-up. The InfinityLab flex bench MS allows you to keep your InfinityLab LC/MSD series and Ultivo LC/TQ systems in one stack with easy access to all areas. It improves lab environments with an integrated solution for waste management and a quiet cover to reduce pump noise.
Specifiche
  • Descrizione:
    Power cord cable assembly, 18-AWG, 3-conductor
  • Seq:
    02