Graphite tubes for PerkinElmer
Graphite tube, pyrocoated, HGA, with advanced platform, HGA 900/850/800/700/600/300 furnaces
HEWL8003-0954
These graphite platforms and tubes for PerkinElmer graphite furnace AA systems are quality tested to ensure every batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance, and lifetime.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
Use partitioned tubes for general-purpose work and for the best detection limits with samples that have low background. These are also ideal when measuring organic solutions, or samples with low viscosity that may spread throughout the tube, potentially degrading precision.
Instruction guides and how-to videos available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
- Per:HGA 900/850/800/700/600/300 furnaces
- Descrizione:Graphite tube, pyrocoated, HGA, with advanced platform
- Seq:02